Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026849.2 | NP_081125.2 | 17 - 18 | 1726 | 81 | |
GenBank mRNA | AK034867.1 | - | 16 - 17 | 1587 | 81 | |
BC012436.1 | - | 15 - 16 | 1254 | 81 | ||
BC018294.1 | - | 17 - 18 | 1676 | 4021243 | 81 | |
DV653632.1 | - | 5 - 6 | 470 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |