Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpusd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033204.1 | NP_001028376.1 | 7 - 8 | 724 | 94 | |
GenBank mRNA | AK076749.1 | - | 7 - 8 | 725 | 94 | |
BC038081.1 | - | 7 - 8 | 665 | 94 | ||
BC116825.1 | - | 7 - 8 | 724 | 40126144 | 94 | |
BC116827.1 | - | 7 - 8 | 724 | 40126146 | 94 | |
BC144734.1 | - | 6 - 7 | 802 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |