Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122892.1 | NP_001116364.1 | 1 - 2 | 388 | 82 | |
NM_001122893.1 | NP_001116365.1 | 1 - 2 | 388 | 82 | ||
NM_008054.2 | NP_032080.2 | 1 - 2 | 388 | 82 | ||
GenBank mRNA | AK156584.1 | - | 1 - 2 | 390 | 82 | |
AK171646.1 | - | 1 - 2 | 390 | 82 | ||
BC092217.1 | - | 1 - 2 | 384 | 6403711 | 82 | |
BQ714273.1 | - | 390 | 82 | |||
M27266.1 | - | 1 - 2 | 100 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |