Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009622.1 | NP_033752.1 | 7 - 8 | 1555 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |