Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400027792.mm.1 |
TC0400003423.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400027792.mm.1 |
NM_001270435.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027792.mm.1 |
NM_001270446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027792.mm.1 |
NM_028506.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027792.mm.1 |
NM_029286.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027791.mm.1 |
TC0400003423.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400027791.mm.1 |
NM_001270435.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027791.mm.1 |
NM_001270446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027791.mm.1 |
NM_028506.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027791.mm.1 |
NM_029286.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027794.mm.1 |
TC0400003423.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400027794.mm.1 |
NM_001270435.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027794.mm.1 |
NM_001270446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027794.mm.1 |
NM_028506.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027794.mm.1 |
NM_029286.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027795.mm.1 |
TC0400003423.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400027795.mm.1 |
NM_001270435.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027795.mm.1 |
NM_001270446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027795.mm.1 |
NM_028506.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027795.mm.1 |
NM_029286.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027798.mm.1 |
TC0400003423.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400027798.mm.1 |
NM_001270446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027797.mm.1 |
TC0400003423.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400027797.mm.1 |
NM_001270446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027793.mm.1 |
TC0400003423.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400027793.mm.1 |
NM_001270435.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027793.mm.1 |
NM_001270446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027793.mm.1 |
NM_028506.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027793.mm.1 |
NM_029286.2 |
Refseq |