Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170333.1 | NP_001163804.1 | 3 - 4 | 574 | 70 | |
GenBank mRNA | BC006623.1 | - | 3 - 4 | 423 | 3968187 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |