Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppil6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028430.1 | NP_082706.1 | 4 - 5 | 533 | 145 | |
GenBank mRNA | AI463820.1 | - | 2 - 3 | 168 | 145 | |
AK013818.1 | - | 4 - 5 | 496 | 145 | ||
BC147756.1 | - | 4 - 5 | 529 | 9056705 | 145 | |
BC147757.1 | - | 4 - 5 | 529 | 9056706 | 145 | |
CA466046.1 | - | 4 - 5 | 515 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |