Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286018.1 | NP_001272947.1 | 4 - 5 | 307 | 79 | |
GenBank mRNA | AK170499.1 | - | 4 - 5 | 308 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |