Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133923.6 | NP_598684.4 | 6 - 7 | 1010 | 74 | |
GenBank mRNA | AK080321.1 | - | 2 - 3 | 424 | 74 | |
AM690746.1 | - | 6 - 7 | 1010 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |