Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsun4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028142.4 | NP_082418.1 | 1 - 2 | 194 | 62 | |
GenBank mRNA | AK012994.1 | - | 1 - 2 | 195 | 62 | |
BC024628.1 | - | 1 - 2 | 114 | 4165638 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |