Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vgll4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177683.2 | NP_808351.1 | 1 - 2 | 437 | 109 | |
GenBank mRNA | AK144447.1 | - | 1 - 2 | 479 | 109 | |
BC048841.1 | - | 1 - 2 | 437 | 5008915 | 109 | |
BC060305.1 | - | 1 - 2 | 450 | 3586396 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |