Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdtc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199306.4 | NP_955010.1 | 5 - 6 | 714 | 96 | |
GenBank mRNA | AK131700.1 | - | 7 - 8 | 874 | 96 | |
AK172431.1 | - | 5 - 6 | 714 | 97 | ||
BC048824.1 | - | 5 - 6 | 508 | 4188979 | 96 | |
BC057107.1 | - | 4 - 5 | 357 | 5697877 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |