Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdtc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199306.4 | NP_955010.1 | 6 - 7 | 900 | 66 | |
GenBank mRNA | AK131700.1 | - | 8 - 9 | 1060 | 66 | |
BC048824.1 | - | 6 - 7 | 694 | 4188979 | 66 | |
BC057107.1 | - | 5 - 6 | 543 | 5697877 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |