Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270435.1 | NP_001257364.1 | 9 - 10 | 1595 | 92 | |
NM_001270446.1 | NP_001257375.1 | 4 - 5 | 851 | 92 | ||
NM_028506.1 | NP_082782.1 | 5 - 6 | 858 | 92 | ||
NM_029286.2 | NP_083562.1 | 9 - 10 | 1610 | 92 | ||
GenBank mRNA | AK049010.1 | - | 4 - 5 | 751 | 92 | |
AK078362.1 | - | 9 - 10 | 1537 | 92 | ||
AK163185.1 | - | 9 - 10 | 1536 | 92 | ||
AK224940.1 | - | 5 - 6 | 787 | 92 | ||
AK224959.1 | - | 4 - 5 | 759 | 92 | ||
BC114436.1 | - | 9 - 10 | 1537 | 40059415 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |