Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Echdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025855.4 | NP_080131.4 | 1 - 2 | 233 | 128 | |
GenBank mRNA | AK006444.1 | - | 1 - 2 | 110 | 128 | |
AK016260.1 | - | 1 - 2 | 175 | 128 | ||
AK166589.1 | - | 1 - 2 | 155 | 128 | ||
AK166634.1 | - | 1 - 2 | 155 | 128 | ||
AK166660.1 | - | 1 - 2 | 155 | 128 | ||
BC066183.1 | - | 1 - 2 | 98 | 30473683 | 128 | |
CJ051963.1 | - | 1 - 2 | 234 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |