Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023906.3 | NP_076395.2 | 6 - 7 | 960 | 73 | |
GenBank mRNA | AF155354.1 | - | 5 - 6 | 775 | 73 | |
AK010315.1 | - | 6 - 7 | 896 | 73 | ||
AK028489.1 | - | 7 - 8 | 1066 | 73 | ||
AK038086.1 | - | 6 - 7 | 941 | 73 | ||
AK143580.1 | - | 6 - 7 | 895 | 73 | ||
BC023086.1 | - | 6 - 7 | 894 | 4947610 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |