Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199351.1 | NP_001186280.1 | 8 - 9 | 914 | 81 | |
NM_001199352.1 | NP_001186281.1 | 8 - 9 | 778 | 81 | ||
NM_012040.3 | NP_036170.1 | 8 - 9 | 854 | 81 | ||
GenBank mRNA | AB023027.1 | - | 7 - 8 | 724 | 81 | |
AF181984.1 | - | 8 - 9 | 828 | 81 | ||
AK134665.1 | - | 8 - 9 | 785 | 81 | ||
AK158755.1 | - | 8 - 9 | 779 | 81 | ||
BC051996.1 | - | 8 - 9 | 761 | 81 | ||
BC055891.1 | - | 8 - 9 | 754 | 4208888 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |