Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028195.3 | NP_082471.2 | 1 - 2 | 178 | 81 | |
GenBank mRNA | AF079972.1 | - | 1 - 2 | 109 | 81 | |
AK030956.1 | - | 1 - 2 | 76 | 81 | ||
AK036679.1 | - | 1 - 2 | 84 | 81 | ||
AK152306.1 | - | 1 - 2 | 84 | 81 | ||
AK152909.1 | - | 1 - 2 | 84 | 81 | ||
AK157026.1 | - | 1 - 2 | 84 | 81 | ||
AK220198.1 | - | 1 - 2 | 107 | 81 | ||
BC018505.1 | - | 1 - 2 | 67 | 81 | ||
BC139054.1 | - | 1 - 2 | 151 | 8862076 | 81 | |
BC139056.1 | - | 1 - 2 | 151 | 8862078 | 81 | |
BC145528.1 | - | 1 - 2 | 151 | 81 | ||
BY161747.1 | - | 1 - 2 | 182 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |