Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028481.1 | NP_082757.1 | 21 - 22 | 3066 | 125 | |
GenBank mRNA | AK016519.1 | - | 21 - 22 | 3060 | 125 | |
BC082598.1 | - | 21 - 22 | 3066 | 30641632 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |