Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fra10ac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081075.1 | NP_001074544.1 | 11 - 12 | 950 | 95 | |
GenBank mRNA | AK077572.1 | - | 11 - 12 | 923 | 95 | |
AK172241.1 | - | 11 - 12 | 957 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |