Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fra10ac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081075.1 | NP_001074544.1 | 7 - 8 | 633 | 140 | |
GenBank mRNA | AK077572.1 | - | 7 - 8 | 606 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |