Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tctn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026486.3 | NP_080762.1 | 13 - 14 | 1619 | 70 | |
GenBank mRNA | AK008223.1 | - | 2 - 3 | 143 | 70 | |
AK014476.1 | - | 13 - 14 | 1585 | 70 | ||
AK031829.1 | - | 13 - 14 | 1620 | 70 | ||
AK032378.1 | - | 12 - 13 | 1704 | 70 | ||
AK162182.1 | - | 1377 | 70 | |||
BC058375.1 | - | 13 - 14 | 1482 | 6828178 | 70 | |
DQ278869.1 | - | 13 - 14 | 1617 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |