Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028031.3 | NP_082307.1 | 11 - 12 | 1336 | 87 | |
GenBank mRNA | AK010382.1 | - | 11 - 12 | 1336 | 87 | |
AK049640.1 | - | 10 - 11 | 1165 | 87 | ||
AK145855.1 | - | 11 - 12 | 1256 | 87 | ||
AK146196.1 | - | 11 - 12 | 1308 | 87 | ||
BC019493.1 | - | 11 - 12 | 1255 | 87 | ||
BC046599.1 | - | 11 - 12 | 1255 | 6311721 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |