Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sppl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029012.2 | NP_083288.2 | 1 - 2 | 98 | 151 | |
GenBank mRNA | AK014709.1 | - | 1 - 2 | 436 | 151 | |
BC086646.1 | - | 1 - 2 | 463 | 6817386 | 151 | |
BU613738.1 | - | 1 - 2 | 211 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |