Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sppl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029012.2 | NP_083288.2 | 7 - 8 | 682 | 82 | |
GenBank mRNA | AK014709.1 | - | 7 - 8 | 1020 | 82 | |
BC023131.1 | - | 3 - 4 | 297 | 82 | ||
BC086646.1 | - | 7 - 8 | 1047 | 6817386 | 82 | |
CD806412.1 | - | 3 - 4 | 299 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |