Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028208.1 | NP_082484.1 | 3 - 4 | 382 | 64 | |
GenBank mRNA | AK006988.1 | - | 3 - 4 | 472 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |