Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027708.1 | NP_081984.1 | 6 - 7 | 1051 | 66 | |
GenBank mRNA | AK016870.1 | - | 6 - 7 | 1052 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |