Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012001.2 | NP_036131.1 | 4 - 5 | 585 | 101 | |
GenBank mRNA | AF071314.1 | - | 4 - 5 | 454 | 101 | |
AK003752.1 | - | 4 - 5 | 447 | 101 | ||
AK009955.1 | - | 4 - 5 | 457 | 101 | ||
BC116800.1 | - | 4 - 5 | 439 | 40126119 | 101 | |
BC116826.1 | - | 4 - 5 | 439 | 40126145 | 101 | |
CX203220.1 | - | 4 - 5 | 587 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |