Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Opn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128599.1 | NP_001122071.1 | 7 - 8 | 1251 | 91 | |
NM_013887.2 | NP_038915.1 | 7 - 8 | 1251 | 91 | ||
GenBank mRNA | AF147789.1 | - | 7 - 8 | 1251 | 91 | |
BC139827.1 | - | 7 - 8 | 1192 | 40142922 | 91 | |
DQ278263.1 | - | 3 - 4 | 414 | 91 | ||
EU303117.1 | - | 7 - 8 | 1067 | 91 | ||
EU303118.1 | - | 7 - 8 | 1067 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |