Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024917.1 | NP_001020088.1 | 13 - 14 | 4630 | 113 | |
GenBank mRNA | AK136182.1 | - | 3 - 4 | 1776 | 113 | |
AK173167.1 | - | 11 - 12 | 4341 | 113 | ||
BC046458.1 | - | 4 - 5 | 2610 | 113 | ||
BC054549.1 | - | 13 - 14 | 4630 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |