Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh18a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019698.2 | NP_062672.2 | 8 - 9 | 1056 | 114 | |
NM_153554.2 | NP_705782.2 | 8 - 9 | 1050 | 114 | ||
GenBank mRNA | AF056573.1 | - | 8 - 9 | 961 | 114 | |
AF056574.1 | - | 8 - 9 | 955 | 114 | ||
AK134148.1 | - | 8 - 9 | 1016 | 114 | ||
AK147711.1 | - | 8 - 9 | 976 | 114 | ||
AK151072.1 | - | 8 - 9 | 978 | 114 | ||
AK153946.1 | - | 8 - 9 | 977 | 114 | ||
AK166459.1 | - | 8 - 9 | 942 | 114 | ||
AK166494.1 | - | 8 - 9 | 943 | 114 | ||
AK168905.1 | - | 8 - 9 | 976 | 114 | ||
BC033427.1 | - | 8 - 9 | 987 | 5009115 | 114 | |
BC037699.1 | - | 8 - 9 | 935 | 4188900 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |