Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phactr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007154.3 | NP_001007155.1 | 4 - 5 | 528 | 66 | |
NM_001177789.1 | NP_001171260.1 | 4 - 5 | 746 | 66 | ||
NM_001177790.1 | NP_001171261.1 | 4 - 5 | 746 | 66 | ||
NM_001177791.1 | NP_001171262.1 | 4 - 5 | 580 | 66 | ||
NM_028806.2 | NP_083082.1 | 4 - 5 | 985 | 66 | ||
GenBank mRNA | AJ632220.1 | - | 4 - 5 | 543 | 66 | |
AJ632221.1 | - | 4 - 5 | 581 | 66 | ||
AJ632222.1 | - | 4 - 5 | 528 | 66 | ||
AJ632223.1 | - | 3 - 4 | 418 | 66 | ||
AK005136.1 | - | 4 - 5 | 550 | 66 | ||
AK032242.1 | - | 4 - 5 | 752 | 66 | ||
AK038449.1 | - | 4 - 5 | 496 | 66 | ||
AK039213.1 | - | 4 - 5 | 535 | 66 | ||
AK135144.1 | - | 4 - 5 | 986 | 66 | ||
AK220554.1 | - | 4 - 5 | 746 | 66 | ||
AY501387.1 | - | 4 - 5 | 692 | 66 | ||
BC058621.1 | - | 4 - 5 | 901 | 5708186 | 66 | |
BC059869.1 | - | 4 - 5 | 764 | 6809310 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |