Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030703.2 | NP_109628.1 | 1 - 2 | 452 | 83 | |
GenBank mRNA | AB021969.1 | - | 1 - 2 | 429 | 83 | |
AF326477.1 | - | 1 - 2 | 452 | 83 | ||
BC014692.1 | - | 1 - 2 | 420 | 4211169 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |