Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bace2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019517.4 | NP_062390.3 | 2 - 3 | 466 | 76 | |
GenBank mRNA | AF216310.1 | - | 2 - 3 | 417 | 76 | |
AK052309.1 | - | 2 - 3 | 453 | 76 | ||
AK078770.1 | - | 2 - 3 | 433 | 76 | ||
AK165036.1 | - | 2 - 3 | 402 | 76 | ||
BC120773.1 | - | 2 - 3 | 432 | 40129696 | 76 | |
BC131947.1 | - | 2 - 3 | 432 | 40130224 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |