Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016786.3 | NP_058066.2 | 2 - 3 | 384 | 140 | |
GenBank mRNA | AB011081.1 | - | 2 - 3 | 322 | 140 | |
AK018366.1 | - | 2 - 3 | 358 | 140 | ||
AK035371.1 | - | 2 - 3 | 281 | 140 | ||
AK036068.1 | - | 2 - 3 | 325 | 140 | ||
AK048194.1 | - | 2 - 3 | 375 | 140 | ||
AK139564.1 | - | 2 - 3 | 401 | 140 | ||
AK160728.1 | - | 2 - 3 | 344 | 140 | ||
AK162142.1 | - | 2 - 3 | 325 | 140 | ||
AK166647.1 | - | 2 - 3 | 293 | 140 | ||
BC002013.1 | - | 2 - 3 | 232 | 3496393 | 140 | |
BC085311.1 | - | 2 - 3 | 310 | 30148009 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |