Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009788.4 | NP_033918.1 | 8 - 9 | 684 | 108 | |
GenBank mRNA | AK002635.1 | - | 8 - 9 | 686 | 108 | |
AK005081.1 | - | 8 - 9 | 683 | 108 | ||
AK005243.1 | - | 8 - 9 | 684 | 108 | ||
AK028050.1 | - | 8 - 9 | 685 | 108 | ||
AK028327.1 | - | 8 - 9 | 683 | 108 | ||
AK048517.1 | - | 8 - 9 | 685 | 108 | ||
AK077499.1 | - | 8 - 9 | 683 | 108 | ||
AK078358.1 | - | 8 - 9 | 682 | 108 | ||
BC016421.1 | - | 8 - 9 | 615 | 4527000 | 108 | |
M21531.1 | - | 8 - 9 | 664 | 108 | ||
M23663.1 | - | 5 - 6 | 246 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |