Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tkt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009388.6 | NP_033414.1 | 8 - 9 | 1248 | 92 | |
GenBank mRNA | AF195533.1 | - | 7 - 8 | 914 | 92 | |
AK002627.1 | - | 8 - 9 | 1201 | 92 | ||
AK012794.1 | - | 8 - 9 | 1203 | 92 | ||
AK030446.1 | - | 8 - 9 | 1231 | 92 | ||
AK140965.1 | - | 8 - 9 | 1232 | 92 | ||
AK144146.1 | - | 8 - 9 | 1203 | 92 | ||
AK145677.1 | - | 8 - 9 | 1201 | 92 | ||
AK146157.1 | - | 8 - 9 | 1201 | 92 | ||
AK150139.1 | - | 8 - 9 | 1201 | 92 | ||
AK150769.1 | - | 8 - 9 | 1197 | 92 | ||
AK150844.1 | - | 8 - 9 | 1197 | 92 | ||
AK150856.1 | - | 8 - 9 | 1197 | 92 | ||
AK150901.1 | - | 8 - 9 | 1200 | 92 | ||
AK151916.1 | - | 8 - 9 | 1196 | 92 | ||
AK151969.1 | - | 8 - 9 | 1196 | 92 | ||
AK152245.1 | - | 8 - 9 | 1196 | 92 | ||
AK152460.1 | - | 8 - 9 | 1217 | 92 | ||
AK152886.1 | - | 8 - 9 | 1201 | 92 | ||
AK159922.1 | - | 8 - 9 | 1201 | 92 | ||
AK166763.1 | - | 8 - 9 | 1235 | 92 | ||
AK167084.1 | - | 8 - 9 | 1235 | 92 | ||
BC055336.1 | - | 8 - 9 | 1175 | 6413263 | 92 | |
U05809.1 | - | 8 - 9 | 1196 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |