Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301424.1 | NP_001288353.1 | 6 - 7 | 966 | 74 | |
NM_007513.4 | NP_031539.3 | 6 - 7 | 1082 | 74 | ||
GenBank mRNA | AK139342.1 | - | 6 - 7 | 1058 | 74 | |
AK147991.1 | - | 6 - 7 | 991 | 74 | ||
AK163202.1 | - | 6 - 7 | 966 | 74 | ||
BC068234.1 | - | 6 - 7 | 973 | 74 | ||
BC145779.1 | - | 5 - 6 | 887 | 40131082 | 74 | |
BC145781.1 | - | 5 - 6 | 887 | 40131084 | 74 | |
JN226407.1 | - | 4 - 5 | 802 | 74 | ||
JN226408.1 | - | 4 - 5 | 802 | 74 | ||
JN226410.1 | - | 4 - 5 | 802 | 74 | ||
M26687.1 | - | 6 - 7 | 1000 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |