Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277149.1 | NP_001264078.1 | 3 - 4 | 2618 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |