Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmeb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198169.2 | NP_937812.1 | 4 - 5 | 477 | 61 | |
GenBank mRNA | AB221583.1 | - | 3 - 4 | 357 | 61 | |
BC029763.1 | - | 3 - 4 | 300 | 61 | ||
BC059066.1 | - | 4 - 5 | 477 | 6825428 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |