Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153135.3 | NP_694775.1 | 5 - 6 | 1044 | 64 | |
GenBank mRNA | AJ487854.1 | - | 5 - 6 | 1045 | 64 | |
BC137615.1 | - | 5 - 6 | 835 | 8860634 | 64 | |
FJ824810.1 | - | 5 - 6 | 977 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |