Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166493.1 | NP_001159965.1 | 15 - 16 | 2107 | 68 | |
NM_207246.4 | NP_997129.1 | 16 - 17 | 2099 | 68 | ||
GenBank mRNA | AB154824.1 | - | 14 - 15 | 1794 | 68 | |
AK173041.1 | - | 15 - 16 | 1992 | 68 | ||
BC066069.1 | - | 14 - 15 | 1737 | 6851971 | 68 | |
BC066127.1 | - | 16 - 17 | 2024 | 30280332 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |