Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015794.1 | NP_056609.1 | 3 - 4 | 1940 | 75 | |
GenBank mRNA | AF176529.1 | - | 3 - 4 | 660 | 75 | |
AK154497.1 | - | 3 - 4 | 449 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |