Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011629.3 | NP_035759.3 | 12 - 13 | 1684 | 135 | |
GenBank mRNA | AK149374.1 | - | 12 - 13 | 1599 | 135 | |
BC090662.1 | - | 12 - 13 | 1660 | 30682075 | 135 | |
BC094580.1 | - | 12 - 13 | 1602 | 30602939 | 135 | |
L26957.1 | - | 11 - 12 | 1493 | 135 | ||
U28265.1 | - | 11 - 12 | 1493 | 135 | ||
U30482.1 | - | 11 - 12 | 1493 | 135 | ||
Y11436.1 | - | 11 - 12 | 2132 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Nuclear Receptor Mammalian Gene Collection Mus musculus Probe spans exons |