Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003919.1 | NP_001003919.1 | 12 - 13 | 1456 | 76 | |
GenBank mRNA | AK049419.1 | - | 12 - 13 | 1462 | 76 | |
AK076357.1 | - | 13 - 14 | 1486 | 76 | ||
AK152868.1 | - | 12 - 13 | 1444 | 76 | ||
BC079656.1 | - | 12 - 13 | 1456 | 6854039 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |