Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026553.4 | NP_080829.1 | 3 - 4 | 532 | 67 | |
GenBank mRNA | AK077329.1 | - | 3 - 4 | 518 | 67 | |
AK160409.1 | - | 3 - 4 | 485 | 67 | ||
BC011117.1 | - | 3 - 4 | 474 | 4190535 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |