Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amotl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019764.2 | NP_062738.2 | 3 - 4 | 1195 | 87 | |
GenBank mRNA | AF175967.1 | - | 3 - 4 | 478 | 87 | |
AF175968.1 | - | 1 - 2 | 1335 | 87 | ||
AK040912.1 | - | 3 - 4 | 1276 | 87 | ||
AK077535.1 | - | 3 - 4 | 1191 | 87 | ||
AK141348.1 | - | 3 - 4 | 1195 | 87 | ||
AK164279.1 | - | 3 - 4 | 1264 | 87 | ||
AK164819.1 | - | 3 - 4 | 1195 | 87 | ||
AK169419.1 | - | 3 - 4 | 1195 | 87 | ||
AX763686.1 | - | 2 - 3 | 1034 | 87 | ||
BC027824.1 | - | 3 - 4 | 1165 | 5323888 | 87 | |
BC035201.1 | - | 3 - 4 | 1177 | 87 | ||
BC108411.1 | - | 3 - 4 | 1229 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |