Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnd3c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033424.3 | NP_001028596.2 | 2 - 3 | 756 | 68 | |
GenBank mRNA | AK030291.1 | - | 2 - 3 | 760 | 68 | |
AK146031.1 | - | 2 - 3 | 757 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |