Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glipr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028608.3 | NP_082884.1 | 1 - 2 | 411 | 63 | |
GenBank mRNA | AK010768.1 | - | 1 - 2 | 343 | 63 | |
AK137557.1 | - | 1 - 2 | 358 | 63 | ||
AK170051.1 | - | 1 - 2 | 358 | 63 | ||
BC025083.1 | - | 1 - 2 | 335 | 3711757 | 63 | |
BC098231.1 | - | 1 - 2 | 385 | 3325823 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |