Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164495.1 | NP_001157967.1 | 4 - 5 | 714 | 68 | |
NM_013722.3 | NP_038750.2 | 4 - 5 | 714 | 68 | ||
GenBank mRNA | AF192754.1 | - | 4 - 5 | 659 | 68 | |
AF498252.1 | - | 3 - 4 | 461 | 68 | ||
AF498253.1 | - | 3 - 4 | 461 | 68 | ||
AK043854.1 | - | 4 - 5 | 715 | 68 | ||
AK048643.1 | - | 4 - 5 | 752 | 68 | ||
AK080557.1 | - | 4 - 5 | 675 | 68 | ||
AK082445.1 | - | 4 - 5 | 752 | 68 | ||
AK084032.1 | - | 4 - 5 | 700 | 68 | ||
AK086955.1 | - | 661 | 68 | |||
AK087342.1 | - | 5 - 6 | 790 | 68 | ||
BC107397.2 | - | 3 - 4 | 580 | 40058225 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |